M6942
Monoclonal Anti-Minor Vault Protein 193 antibody produced in mouse
250 μg/mL, clone p193-10, tissue culture supernatant
Synonym(s):
Anti-VPARP, Anti-p193
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
tissue culture supernatant
antibody product type
primary antibodies
clone
p193-10, monoclonal
species reactivity
human
concentration
250 μg/mL
technique(s)
immunocytochemistry: 1:10-1:20 using 4% paraformaldehyde-fixed cytospin preparations
immunohistochemistry (frozen sections): 1:10-1:20
western blot (chemiluminescent): 1:10-1:20
isotype
IgG2aκ
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PARP4(143)
Looking for similar products? Visit Product Comparison Guide
General description
Specificity
Immunogen
Biochem/physiol Actions
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service