Skip to Content
Merck
All Photos(1)

Documents

M6877

Sigma-Aldrich

α-Methyl-DL-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white

SMILES string

CC(N)(CO)C(O)=O

InChI

1S/C4H9NO3/c1-4(5,2-6)3(7)8/h6H,2,5H2,1H3,(H,7,8)

InChI key

CDUUKBXTEOFITR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Methyl-DL-serine is an amino acid derivative.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I S Mendelson
Journal of mental deficiency research, 26 (Pt 2), 107-110 (1982-06-01)
An inherited defect in the glycine cleavage enzyme results in the condition of neonatal glycine encephalopathy which has not responded to the current innovative methods of therapy. A re-examination of the enzyme structure and metabolic pathways, leads us to recommend
H Mickos et al.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989), 46(10), 989-993 (1992-10-01)
The three-dimensional structure of the RGD-adhesion sequence has been studied previously by means of linear and cyclic peptides. These peptides show widely differing affinities to integrins, ascribed to a strong dependence on steric factors in the receptor recognition. Insertion of
Francisco Corzana et al.
Chemical communications (Cambridge, England), 47(18), 5319-5321 (2011-04-01)
A novel Tn antigen mimic, in which the natural underlying amino acid has been replaced by the non-natural α-methylserine analogue, is reported. This derivative exhibits a similar affinity for a natural lectin as for the natural Tn and retains the
Ewa Zabłotna et al.
Biochemical and biophysical research communications, 340(3), 823-828 (2005-12-29)
In many complexes formed by serine proteinases and their inhibitors, the hydroxyl group provided by water molecule or by the inhibitor Ser residue is located close to the inhibitor P1-P1' reactive site. In order to investigate the role of this
E Altmann et al.
International journal of peptide and protein research, 32(5), 344-351 (1988-11-01)
The conformational behaviour of host-guest peptides of the type Ac-Ala-Xxx-Ala-Ala-Xxx-Ala-Ala-Xxx-Ala-Ala-NH-PEGM (Xxx = alpha-aminoisobutyric acid (Aib), (S)-2-ethylalanine ((S)-Iva), (S)-2-methylserine ((S)-alpha-MeSer)) has been studied by CD spectroscopy in CF3CH2OH, CH3OH, and water and by i.r. spectroscopy in CHCl3 and in the solid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service