Skip to Content
Merck
All Photos(1)

Key Documents

M6320

Sigma-Aldrich

Monoclonal Anti-MAGE-1 antibody produced in mouse

~2 mg/mL, clone MA454, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Melanoma-associatiated antigen 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$727.00

$727.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$727.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$727.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MA454, monoclonal

form

buffered aqueous solution

mol wt

antigen ~46 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 5-10 μg/mL using total cell extract of SKML37 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGEA1(4100)

General description

Melanoma-associatiated antigen 1 (MAGE-1) is part of the CG (cancer-germline) group of genes and is a tumor-specific antigen.[1][2] MAGE-1 is a member of the MAGE-A family. MAGE-1 gene is localized on human chromosome Xq28.[3] Monoclonal Anti-MAGE-1 (mouse IgG1 isotype) is derived from the hybridoma MA454 produced by the fusion of mouse myeloma cells (SP2/0 cells) and splenocytes from mice immunized with human recombinant MAGE-1.

Specificity

The antibody does not recognize MAGE-2 and MAGE-3.[4][5]

Immunogen

recombinant human MAGE-1.

Application

Monoclonal Anti-MAGE-1 antibody produced in mouse has been used in:
  • ELISA
  • immunoblotting
  • immunohistochemistry

Biochem/physiol Actions

The melanoma antigen genes (MAGE) might play a key role in neural development and apoptosis. Expression of melanoma-associated antigen 1 (MAGE-1) in neoplastic cells depends on the binding of the transcription factor Ets to non-methylated cis regulatory sequences, located in its promoter. MAGE-1 expression is mediatedby the B and B′ domains of MAGE-A1 gene. Expression of MAGE-1A was induced in non MAGE-A1 expressing tumor cells once treated with demethylating reagent, 5-aza-2-(5DC).[6] Melanoma-associatiated antigen 1 (MAGE-1) has been shown to be expressed in different types of cancers.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The MAGE Proteins: Emerging Roles in Cell
Cycle Progression, Apoptosis, and
Neurogenetic Disease
Barker PA and Salehi A
Journal of Neuroscience Research (2002)
Clinical Significance of Melanoma Antigen-Encoding Gene-1 (MAGE-1) Expression and its Correlation with Poor Prognosis in Differentiated Advanced Gastric Cancer
Ogata MDK, et al.
Annals of Surgical Oncology (2011)
Y T Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(3), 1004-1008 (1994-02-01)
The human MAGE-1 gene encodes a melanoma peptide antigen recognized by autologous cytotoxic T lymphocytes. To produce antibodies against the MAGE-1 gene product, several approaches were taken. Three oligopeptides were synthesized based on predicted MAGE-1 amino acid sequences and were
Madhav V Dhodapkar et al.
Cancer immunity, 3, 9-9 (2003-07-24)
Cancer/testis (CT) antigens are expressed in several malignant tumors, but not in normal tissues except for testicular germ cells. The expression of CT antigenic proteins in malignant gammopathies has not been characterized. We examined the expression of a panel of
Jiang Xiao et al.
World journal of gastroenterology, 10(13), 1849-1853 (2004-06-29)
To date, dozens of melanoma-associated antigens (MAGEs) have been identified and classified into 2 subgroups, I and II. Subgroup I consists of antigens which expression is generally restricted to tumor or germ cells, also named as cancer/testis (CT) antigen. Proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service