M5192
Anti-Matrix Metalloproteinase-26, Propeptide Region antibody produced in rabbit
~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-MMP-26
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous glycerol solution
species reactivity
human
concentration
~1 mg/mL
technique(s)
western blot: 1:1,000
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MMP26(56547)
General description
Matrix metalloproteinase-26 (MMP-26) is also known as matrilysin-2 and endometase. It is expressed in normal tissues and also in human carcinoma cells. MMP-26 possesses a propeptide domain, a signal peptide and a catalytic domain but does not have the hemopexin-like domain which is common to other members of its family. The MMP-26 gene is localized to human chromosome 11p15.3.
Specificity
By immunoblotting against the reduced protein, the antibody identifies a band at 30 kDa (zymogen).
Immunogen
synthetic peptide corresponding to the propeptide domain of human matrix metalloproteinase-26.
Biochem/physiol Actions
Matrix metalloproteinase-26 (MMP-26) degrades fibronectin, type IV collagen and activated pro-metalloproteinase-9. MMP-26 also aids in development of tumors.
Physical form
Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of Cancer, 4(4), 296-303 (2013-04-10)
Human endometase/matrilysin-2/matrix metalloproteinase-26 (MMP-26) is an endopeptidase mostly produced by human carcinoma cells. While MMPs are thought to regulate the dynamics of extracellular matrix turnover, new evidence shows that these enzymes may play a critical regulatory role in inflammation. To
The Biochemical journal, 356(Pt 3), 705-718 (2001-06-08)
Identification of expanding roles for matrix metalloproteinases (MMPs) in complex regulatory processes of tissue remodelling has stimulated the search for genes encoding proteinases with unique functions, regulation and expression patterns. By using a novel cloning strategy, we identified three previously
Molecular medicine reports, 4(6), 1201-1209 (2011-08-02)
Matrix metalloproteinase 26 (MMP-26) is a novel member of the matrix metalloproteinase (MMP) family and is widely expressed in cancer cells of epithelial origin. MMP-26 has been shown to contribute to tumor development and to the restoration of tissue injury.
Experimental and molecular pathology, 92(1), 59-63 (2011-09-29)
Matrix metalloproteinase (MMP) regulation and expression is important in the aging/degenerating human intervertebral disc. MMP-26 (also known as matrilysin-2 or endometase) is a newly discovered MMP which degrades type IV collagen, fibronectin, fibrinogen, vitronectin, denatured collagen types I-IV, insulin-like growth
Oncology reports, 30(2), 751-756 (2013-06-12)
The human matrix metalloproteinase (MMP)-26, also called matrilysin-2 or endometase, has been isolated as a matrilysin (MMP-7) homolog. Several reports describe that MMP-26 may be related to the development of endometrial carcinomas. Total RNAs were isolated from 51 normal endometrial
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service