Skip to Content
Merck
All Photos(1)

Key Documents

M5191

Sigma-Aldrich

Anti-Matrix Metalloproteinase-28, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Epilysin, Anti-MMP-28

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$741.00

$741.00


Check Cart for Availability


Select a Size

Change View
100 μG
$741.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$741.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP28(79148)

General description

Matrix metalloproteinase-28 (MMP-28) is expressed in normal tissues like the epidermis of the skin, brain, lungs and intestine. It possesses an activation sequence and a pro-domain. The gene encoding it is localized on human chromosome 17q11.2 and has eight exons.

Specificity

By immunoblotting against the reduced protein, the antibody identifies bands at 62 kDa and 58 kDa, as well as breakdown products at 50 kDa, 48 kDa, and 46 kDa. At higher antibody concentrations, the antibody also detects a 70 kDa band in rat and mouse tissues.

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-28.

Biochem/physiol Actions

Matrix metalloproteinase-28 (MMP-28) may have a role in modulating matrix composition. It is an endopeptidase which degrades casein and mediates epithelial to mesenchymal transition. MMP-28 aids in progression of tumors and is overexpressed in gastric carcinomas.

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mRNA expression of MMP-28 (Epilysin) in gingival tissues of chronic and aggressive periodontitis patients: a reverse transcriptase PCR study.
Padmavati P
Disease Markers, 35(2), 113-118 (2013)
MMP28 and macrophage polarization: orchestrating the attack of the mac.
Ye Gan et al.
Journal of leukocyte biology, 95(1), 1-3 (2014-01-05)
Helen E Gruber et al.
Arthritis research & therapy, 11(6), R184-R184 (2009-12-17)
The regulation and elevation in expression of the catabolic matrix metalloproteinases (MMPs) is of high importance in the human intervertebral disc since upregulation of these matrix-degrading enzymes results in matrix destruction associated with disc degeneration. MMP28 (epilysin) is a newly
Tuomas J Heiskanen et al.
Matrix biology : journal of the International Society for Matrix Biology, 28(2), 74-83 (2009-04-22)
Epilysin (MMP-28) is a conserved member of the matrix metalloproteinase (MMP) family. It is expressed in various normal tissues, and induced in wounds and in developing and regenerating nerves. Epilysin induces TGF-beta mediated epithelial to mesenchymal transition, but its other
Marina Klawitter et al.
Journal of negative results in biomedicine, 10, 9-9 (2011-08-02)
MMP28 (epilysin) is a recently discovered member of the MMP (matrix metalloproteinase) family that is, amongst others, expressed in osteoarthritic cartilage and intervertebral disc (IVD) tissue. In this study the hypothesis that increased expression of MMP28 correlates with higher grades

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service