Skip to Content
Merck
All Photos(1)

Key Documents

M5171

Sigma-Aldrich

S-Methyl-L-thiocitrulline acetate salt

≥98% (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$133.00

$133.00


Check Cart for Availability


Select a Size

Change View
10 MG
$133.00

About This Item

Empirical Formula (Hill Notation):
C7H15N3O2S · C2H4O2
CAS Number:
Molecular Weight:
265.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

$133.00


Check Cart for Availability

Assay

≥98% (TLC)

storage temp.

−20°C

SMILES string

CC(O)=O.CSC(=N)NCCC[C@H](N)C(O)=O

InChI

1S/C7H15N3O2S.C2H4O2/c1-13-7(9)10-4-2-3-5(8)6(11)12;1-2(3)4/h5H,2-4,8H2,1H3,(H2,9,10)(H,11,12);1H3,(H,3,4)/t5-;/m0./s1

InChI key

XUKPZPRDNPUAJY-JEDNCBNOSA-N

Biochem/physiol Actions

Potent inhibitor of inducible nitric oxide synthase with preference for constitutive (neuronal) over inducible (endothelial) NOS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Narayanan et al.
Journal of medicinal chemistry, 37(7), 885-887 (1994-04-01)
Nitric oxide synthase catalyzes the NADPH- and O2-dependent conversion of L-arginine to L-citrulline and nitric oxide. L-Thiocitrulline, L-homothiocitrulline, and S-methyl-L-thiocitrulline, novel citrulline analogs, have been synthesized and are shown to be potent inhibitors of both the constitutive brain and the
Liliana M Sanmarco et al.
Nature, 611(7937), 801-809 (2022-10-21)
Genome-wide association studies have identified risk loci linked to inflammatory bowel disease (IBD)1-a complex chronic inflammatory disorder of the gastrointestinal tract. The increasing prevalence of IBD in industrialized countries and the augmented disease risk observed in migrants who move into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service