Skip to Content
Merck
All Photos(1)

Key Documents

M5037

Sigma-Aldrich

2-(Methylamino)propiophenone hydrochloride

Synonym(s):

1-Propanone, 2-(methylamino)-1-phenyl-, hydrochloride, DL-Ephedrone hydrochloride, Methcathinone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO · HCl
CAS Number:
Molecular Weight:
199.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Product M5037 is not currently sold in your country. Contact Technical Service

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

application(s)

forensics and toxicology

SMILES string

Cl[H].CNC(C)C(=O)c1ccccc1

InChI

1S/C10H13NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,11H,1-2H3;1H

InChI key

LTJXNCHDKYZOSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Methylamino)propiophenone hydrochloride has been used as a positive control in forced swimming test.[1]

Biochem/physiol Actions

Potent amphetamine-like stimulant; dimethylpropion metabolite.

Other Notes

Mixture of optical isomers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anxiolytic and antidepressive effects of the homeopathic complex [TM="Homeo-pax"](pre-clinical study)
Vaz AF, et al.
International Journal of High Dilution Research, 8(28), 04-14 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service