Skip to Content
Merck
All Photos(1)

Key Documents

M4154

Sigma-Aldrich

β-N-Methylamino-L-alanine hydrochloride

Synonym(s):

L-BMAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N2O2 · HCl
CAS Number:
Molecular Weight:
154.60
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

CNCC(N)C(O)=O

Biochem/physiol Actions

Neuroexcitotoxic amino acid; may be involved in Guam amyotrophic lateral sclerosis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

András Guseo
Orvosi hetilap, 153(52), 2060-2069 (2012-12-25)
Parkinson's disease is one of the most frequent progressive degenerative disorders with unknown origin of the nervous system. The commutation of the disease on Guam led to the discovery of a neurotoxin which was also found in other continents. This
Elijah W Stommel et al.
Medical hypotheses, 80(2), 142-145 (2012-12-19)
Sporadic amyotrophic lateral sclerosis (sALS) is a fatal neurodegenerative disease with no known cause. There are many clues to suggest an environmental trigger for the disease, including reports of conjugal couples and co-localized employees that developed sALS. On the island
Travis Rush et al.
Neuroreport, 23(4), 216-219 (2012-02-09)
Determination of the environmental factors involved in neurodegenerative diseases has been elusive. Methylmercury and β-N-methylamino-L-alanine (BMAA) have both been implicated in this role. Exposure of primary cortical cultures to these compounds independently induced concentration-dependent neurotoxicity. Importantly, concentrations of BMAA (10-100
Kiyo Mondo et al.
Marine drugs, 10(2), 509-520 (2012-03-14)
Sharks are among the most threatened groups of marine species. Populations are declining globally to support the growing demand for shark fin soup. Sharks are known to bioaccumulate toxins that may pose health risks to consumers of shark products. The
Oskar Karlsson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 130(2), 391-404 (2012-08-09)
The cyanobacterial toxin β-N-methylamino-L-alanine (BMAA) has been proposed to contribute to neurodegenerative disease. We have previously reported a selective uptake of BMAA in the mouse neonatal hippocampus and that exposure during the neonatal period causes learning and memory impairments in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service