Skip to Content
Merck
All Photos(1)

Key Documents

M3656

Sigma-Aldrich

Mephentermine hemisulfate salt

Synonym(s):

N,α,α-Trimethylphenethylamine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H17N · 1/2H2SO4
CAS Number:
Molecular Weight:
212.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

SMILES string

OS(O)(=O)=O.CNC(C)(C)Cc1ccccc1.CNC(C)(C)Cc2ccccc2

Biochem/physiol Actions

α-adrenoceptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mohta et al.
Anaesthesia and intensive care, 37(4), 568-570 (2009-08-18)
This study was designed to determine the minimum effective dose (ED50) of mephentermine for prevention of post-spinal hypotension in women undergoing elective caesarean section. Dixon's up-down method of sequential allocation was used to determine the patient's dose of vasopressor drug.
B Bhattarai et al.
JNMA; journal of the Nepal Medical Association, 49(177), 23-28 (2010-12-25)
Hypotension is common following spinal anesthesia. Various vasopressors have been indicated to prevent it. The study compares three such agents namely phenylephrine, ephedrine and mephentermine. The study included 90 patients undergoing elective and emergency cesarean section who developed hypotension following
Up-down study--is the analysis adequate?
J K Makkar et al.
Anaesthesia and intensive care, 38(1), 208-209 (2010-03-03)
T Murali Krishna et al.
European journal of anaesthesiology, 25(4), 299-306 (2007-09-26)
Intrathecal ketamine produces a short period of analgesia with stable haemodynamics. Midazolam with bupivacaine prolongs the duration of analgesia when administered intrathecally but does not prevent hypotension. The objective of this study was to assess the effect of a combination
M Mori et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(3), 287-299 (1989-03-01)
1. Excretion of urinary and biliary radioactivity, and metabolites of [3H]mephentermine (MP), after i.p. or subcutaneous administration of [3H]MP to male Wistar rats, were determined by preparative t.l.c.-liquid scintillation counting. 2. About 45% of the radioactivity administered i.p. was excreted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service