Skip to Content
Merck
All Photos(1)

Key Documents

M3420

Sigma-Aldrich

Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside

≥90% (TLC)

Synonym(s):

6-O-(N-Heptylcarbamoyl)-methyl-α-D-glucopyranoside, HECAMEG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NO7
CAS Number:
Molecular Weight:
335.39
Beilstein:
3560078
MDL number:
UNSPSC Code:
12161900

description

non-ionic

Assay

≥90% (TLC)

storage temp.

−20°C

SMILES string

CCCCCCCNC(=O)OC[C@H]1O[C@H](OC)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H29NO7/c1-3-4-5-6-7-8-16-15(20)22-9-10-11(17)12(18)13(19)14(21-2)23-10/h10-14,17-19H,3-9H2,1-2H3,(H,16,20)/t10-,11-,12+,13-,14+/m1/s1

InChI key

XPIVOYOQXKNYHA-RGDJUOJXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent with superior properties: dissociates protein aggregates but does not denature enzymes or antigens; high CMC (19.5 mM) allow fast removal by dialysis.

replaced by

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Plusquellec et al.
Analytical biochemistry, 179(1), 145-153 (1989-05-15)
A new surfactant, 6-O-(N-heptylcarbamoyl)-methyl-alpha-D-glucopyranoside (HECAMEG, molar mass 335.38 g), was synthesized by a simple and low cost procedure from methyl-alpha-D-glucopyranoside. This surfactant is characterized by a high solubility in water (even at 0 degree C), ultraviolet light transparency in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service