Skip to Content
Merck
All Photos(1)

Key Documents

M3379

Sigma-Aldrich

DL-Methioninol

≥95%

Synonym(s):

4-Methylmercapto-2-amino-1-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13NOS
CAS Number:
Molecular Weight:
135.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-Methioninol, ≥95%, viscous liquid

Assay

≥95%

form

viscous liquid

color

colorless to faint yellow

application(s)

cell analysis

SMILES string

CSCCC(N)CO

InChI

1S/C5H13NOS/c1-8-3-2-5(6)4-7/h5,7H,2-4,6H2,1H3

InChI key

MIQJGZAEWQQAPN-UHFFFAOYSA-N

Biochem/physiol Actions

Methioninol is a methionine derivative that has been shown to inhibit SDK and TREK-1 channels in COS cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methionine and its derivatives increase bladder excitability by inhibiting stretch-dependent K+ channels.
Baker, S.A. et al.
British Journal of Pharmacology, 53, 1259-1271 (2008)
S A Baker et al.
British journal of pharmacology, 153(6), 1259-1271 (2008-01-22)
During the bladder filling phase, the volume of the urinary bladder increases dramatically, with only minimal increases in intravesical pressure. To accomplish this, the smooth muscle of the bladder wall must remain relaxed during bladder filling. However, the mechanisms responsible
Homar S Barcena et al.
Organic letters, 5(5), 709-711 (2003-02-28)
One-electron reduction inverts the helicity of copper complexes formed from derivatives of S-methylcysteine and methioninol. The change in conformation of the organic ligand is followed in the exciton-coupled circular dichroism (ECCD) spectra of the complexes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service