Skip to Content
Merck
All Photos(1)

Key Documents

M266

Sigma-Aldrich

Anti-Glutamate Receptor NMDAR2C (NR2C) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-GluN2C, Anti-NMDAR2C, Anti-NR2C

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$1,290.00

$1,290.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 μG
$1,290.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$1,290.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen 140 kDa (NR2C subunit)
antigen 180 kDa (NR2A and NR2B bands)

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry (frozen sections): 1:1000
immunoprecipitation (IP): 3 μL using 200 μg of rat cerebellum.
indirect immunofluorescence: 1:1000 using frozen sections
western blot: 1:1000 using 200 μg cell lysate

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIN2C(2905)
mouse ... Grin2c(14813)
rat ... Grin2c(24411)

Immunogen

N-terminal portion of NR2C (amino acids 25-130)

Physical form

Lyophilized from ammonium bicarbonate (5 mM).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane Peineau et al.
Methods in molecular biology (Clifton, N.J.), 1677, 253-263 (2017-10-08)
Proving endogenous GluN presence and functions in microglia require complementary steps to demonstrate (1) that GluN genes are transcripted and translated, (2) their cellular localization, (3) that the GluN are functional, and (4) the role of the functional GluN. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service