M2656
Multiwell plate washer/dispenser manifolds
8 position, straight
Synonym(s):
Multiwell Washer, Plate Washer, Plate Washer Manifolds
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
sterility
non-sterile
color
none
Looking for similar products? Visit Product Comparison Guide
General description
For use with 96 well multiwell plates
- Dispense or aspirate media, or wash, a full row or column at a time
- Luer fitting
- Stainless steel and PTFE construction for easy cleaning and autoclavability
- Optional repeat kit provides continuous operation capability
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service