Skip to Content
Merck
All Photos(1)

Key Documents

M0506

Sigma-Aldrich

N-Methyl-DL-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

SMILES string

CNC(C)C(O)=O

InChI

1S/C4H9NO2/c1-3(5-2)4(6)7/h3,5H,1-2H3,(H,6,7)

InChI key

GDFAOVXKHJXLEI-UHFFFAOYSA-N

Biochem/physiol Actions

N-Methyl-DL-alanine may be used in studies of amino acid transport mechanisms such as trans-stimulation. N-Methylalanine may be used in the construction of peptide and protein analogues for use in drug or antibiotic design and development.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Lakshminarayana et al.
Indian journal of experimental biology, 38(4), 373-378 (2001-02-24)
Spontaneous mutants resistant to methionine sulfoximine (Msx), methyl alanine (Mal) and methyl ammonium chloride (Mac) were derived from A. chroococcum strain A103. Msx and Mal-resistant mutants expressed 1.73 to 10.98% of the fully derepressed nitrogenase activity when grown in Burk's
G E Mann et al.
The Journal of physiology, 416, 485-502 (1989-09-01)
1. Epithelial uptake and efflux of the non-metabolized system A analogue 2-methylaminoisobutyric acid (MeAIB) and L-serine were studied in the isolated perfused rat pancreas using a dual tracer loading and wash-out technique. Uptakes of 2-[14C]MeAIB and L-[3H]serine were measured relative
Renata Perlikowska et al.
Basic & clinical pharmacology & toxicology, 106(2), 106-113 (2009-10-31)
A series of endomorphin-1 (EM-1) and endomorphin-2 (EM-2) analogues, containing non-cyclic amino acids (Ala, D-Ala, beta-Ala, NMeAla, D-NMeAla or Sar) instead of Pro in position 2 was synthesized, where NMeAla = N-methylalanine and Sar = N-methylglycine, sarcosine. The opioid activity
Bang-zhi Zhang et al.
Peptides, 31(4), 568-573 (2010-01-05)
The actinomycin D (AMD) analogs in which the D-valine residues (the second amino acid residue in the cyclic depsipeptide of AMD) and the N-methyl-L-valine residues (the fifth amino acid residue in the cyclic depsipeptide of AMD) were replaced with D-Phe
G Yadid et al.
Neuroscience, 55(4), 1147-1152 (1993-08-01)
[3H]Glycine is actively taken up into bovine isolated adrenal medulla chromaffin cells with the subsequent catecholamine release. [3H]Glycine uptake has two interaction sites based on relative Km measurements. These sites are inherently distinct since the effects of strychnine and temperature

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service