Skip to Content
Merck
All Photos(1)

Key Documents

M041100

Sigma-Aldrich

6-Fluorescein Phosphoramidite

Synonym(s):

6-FAM-phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H58N3O10P
CAS Number:
Molecular Weight:
843.94
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥97% (31P-NMR)
≥97.0% (reversed phase HPLC)

color

solid white to off-white

solubility

acetonitrile: 0.1 M, clear, colorless to faintly yellow (liquid)

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCCNC(=O)c1ccc2C(=O)OC3(c4ccc(OC(=O)C(C)(C)C)cc4Oc5cc(OC(=O)C(C)(C)C)ccc35)c2c1)OCCC#N

InChI

1S/C46H58N3O10P/c1-29(2)49(30(3)4)60(55-25-15-22-47)54-24-14-12-11-13-23-48-40(50)31-16-19-34-37(26-31)46(59-41(34)51)35-20-17-32(56-42(52)44(5,6)7)27-38(35)58-39-28-33(18-21-36(39)46)57-43(53)45(8,9)10/h16-21,26-30H,11-15,23-25H2,1-10H3,(H,48,50)

InChI key

MGPYJVWEJNTXLC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service