Skip to Content
Merck
All Photos(1)

Key Documents

M024030

Sigma-Aldrich

ssH-Linker

configured for ABI

Synonym(s):

9,11-Dioxa-2-aza-10-phosphatridecanoic acid, 10-[bis(1-methylethyl)amino]-13-cyano-, 2-[[(4-methoxyphenyl)diphenylmethyl]amino]ethyl este

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
$218.00

$218.00


Check Cart for Availability


Select a Size

Change View
0.25 G
$218.00

About This Item

Empirical Formula (Hill Notation):
C38H53N4O5P
Molecular Weight:
676.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

$218.00


Check Cart for Availability

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

compatibility

configured for ABI

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(NCCOC(=O)NCCCCCCOP(OCCC#N)N(C(C)C)C(C)C)(c2ccccc2)c3ccccc3

InChI

1S/C38H53N4O5P/c1-31(2)42(32(3)4)48(47-29-16-25-39)46-28-15-7-6-14-26-40-37(43)45-30-27-41-38(33-17-10-8-11-18-33,34-19-12-9-13-20-34)35-21-23-36(44-5)24-22-35/h8-13,17-24,31-32,41H,6-7,14-16,26-30H2,1-5H3,(H,40,43)

InChI key

VTFLXOVDKJUHER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

This product is covered under US Patent Number 7,491,857

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service