Skip to Content
Merck
All Photos(1)

Key Documents

LYSC9001

Sigma-Aldrich

Lys-c (lysyl-endopeptidase), Active

from Achromobacter lyticus, recombinant, expressed in E. coli (206-473aa), tag-free, powder

Synonym(s):

API, Lysyl endopeptidase, Protease I

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$721.00
100 μG
$2,520.00

$721.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$721.00
100 μG
$2,520.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

$721.00


Check Cart for Availability

Request a Bulk Order

biological source

Achromobacter lyticus

Quality Level

recombinant

expressed in E. coli (206-473aa)

grade

Proteomics Grade

form

lyophilized powder

optimum pH

8.5

pH range

9.0-9.5(catalytic)

UniProt accession no.

storage temp.

−20°C

General description

Recombinant tag-free Achromobacter lyticus Lys-c (lysyl-endopeptidase) (206-473aa) was expressed in E.coli cells.

Overview
Lysyl-endopeptidase (Lys-c) was isolated from the Gram-negative soil bacterium Achromobacter lyticus by Msaki et al. The protein hydrolyzes amide and peptide ester bonds at the carboxylic side of lysine and S-aminoethylcysteine residues making it an important tool for enzymatic protein sequencing and Lys-X compound synthesis.

Specificity

This enzyme hydrolyzes amide and peptide ester bonds at the carboxylic side of lysine and S-aminoethylcysteine residues, at a catalytic pH range of 9.0-9.5, catalytic temperature range of 30-37 °C.

Application

The enzyme functions optimally between 30-37 °C and suffers from degradation when subjected to temperatures above 50 °C. Lysyl-endopeptidase retains complete activity after incubation in 4M urea or in 0.1% SDS solution for up to 6 hours at 30 °C.

Packaging

1mg/ml in Plastic

Preparation Note

Catalytic pH range 9.0-9.5. Catalytic temperature range 30-37 °C.

Reconstitution

Reconstitute in sterile water.

Storage and Stability

Store product at -20°C. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles once the protein has been resolubilized in sterile water.

Other Notes

For R&D only.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service