Skip to Content
Merck
All Photos(1)

Key Documents

LPLL001

Sigma-Aldrich

Laminin/Poly-L-Lysine Coating Solution

0.2 μm filtered, BioReagent, suitable for cell culture, Ready-to-use mixture

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$330.00

$330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$330.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

$330.00


Check Cart for Availability

Request a Bulk Order

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

liquid

concentration

1 ×

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Laminin/Poly-L-Lysine Coating Solution is a ready-to-use attachment factors mix solution, prepared in phosphate-buffered saline and is 0.2µm filtered. It is used to coat cell culture flasks, dishes, and multi-well plates and used to promote the attachment, spreading and proliferation of variety of cell types that require ECM-coated surface for adhesion and 2D cell-culture growth. The combination of Poly-L-Lysine and Laminin has been used for optimal attachment and promotion of neurite outgrowth in culture. It is suitable for culturing many different types of Peripheral Nervous System (PNS) and Central Nervous System (CNS) networks and is useful for promoting neural cell attachment and differentiation. Certain cells can digest Poly-L-lysine, in which case the Laminin/Poly-D-Lysine solution can be used (Cat# LPDL001).

Application

Laminin/Poly-L-Lysine Coating Solution can be used in:
  • Attachment and spreading of a variety of cell types
  • Enhancement of neuronal cell attachment to plastic and glass
  • Support of neurite outgrowth
  • Suitable for use with serum-free or reduced -serum cultures

Biochem/physiol Actions

Laminin proteins are integral components of structural scaffolding in animal tissues. Laminin has active domains for collagen binding, cell adhesion, heparin binding, and neurite outgrowth fragment. Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells.

Poly-L-lysine hydrobromide is a nonspecific attachment factor for cells, useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, Poly-L-lysine increases the number of positively charged cell binding sites.

Components

Laminin is a large basement membrane glycoprotein and composed of three different polypeptide chains, termed α, β and γ. The cohesion between these chains is the result of many inter and intrachain disulfide bonds. Together, they cause the molecule to look like a crucifix.

Preparation Note

LPLL001 is a ready to use mixture composed of Laminin and Poly-L-Lysine. It is filtered using 0.2µm filter. Gently mix the Laminin/Poly-L-Lysine Coating Solution a few times to form a homogenous solution before use. For use as a coating solution, cover the entire culture surface area, approximately 0.2-0.25 mL/cm2. Incubate at room temperature for 2-3 hours and carefully aspirate the solution and wash 3 times with sterile PBS before plating cells. The coated tissue culture vessels can be used immediately or stored at 4 °C for up to one week filled with PBS and wrapped in parafilm. Remove PBS only when ready to plate the cells. Do not let the coated plates dry completely

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service