Skip to Content
Merck
All Photos(1)

Key Documents

L9152

Sigma-Aldrich

Anti-Leukemia Inhibitory Factor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

CDF, D factor, DIA, Emfilermin, LIF, differentiation inhibitory activity

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$973.00

$973.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$973.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.43

$973.00


Check Cart for Availability

Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIF(3976)
mouse ... Lif(16878)

Looking for similar products? Visit Product Comparison Guide

Specificity

The antibody neutralizes the biological activity of recombinant mouse LIF and also neutralizes the biologicl activity of recombinant human LIF at a 10-fold higher IgG concentration. Based on ELISA and immunoblotting, this antibody shows <10% cross-reactiity with recombinant human LIF.

Immunogen

purified, E. coli-derived, recombinant mouse LIF.

Application

For Western blot a concentration of 0.1 μg/ml is recommended. For immunohistochemistry, a working dilution of 5-15 μg/ml is recommended. The neutralization dose is typically 80-480 ng/mL in the presence of 3 ng/mL Recombinant Mouse LIF.

Optimal dilutions should be determined by the end user.

Biochem/physiol Actions

Leukemia inhibitory factor (LIF) was initially identified as a factor that inhibited the proliferation and induced the differentiation to macrophages of the murine myeloid leukemic cell line M1. Subsequent to its purification and molecular cloning, LIF was recognized to be a pleiotropic factor with multiple effects on both hematopoietic and non-hematopoietic cells. LIF has overlapping biological functions with OSM, IL-6, IL-11 and CNTF. All these cytokines utilize gp130 as a component in their signal transducing receptor complexes. Mouse LIF cDNA encodes a 203 amino acid residue polypeptide with a 23 amino acid signal peptide that is cleaved to yield a 180 amino acid mature mouse LIF. Native human and mouse LIF are highly glycosylated monomeric proteins. Both human and murine LIF protein sequences have multiple potential N- and O-linked glycosylation sites and six conserved cysteine residues that are involved in three intramolecular disulfide bridges.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.

Preparation Note

Protein A or G purified

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Tsukada et al.
PloS one, 10(6), e0129011-e0129011 (2015-06-05)
Recent studies in rodents suggest that maternal immune activation (MIA) by viral infection is associated with schizophrenia and autism in offspring. Although maternal IL-6 is though t to be a possible mediator relating MIA induced these neuropsychiatric disorders, the mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service