Skip to Content
Merck
All Photos(1)

Key Documents

L8250

Sigma-Aldrich

DL-Leucinamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O · HCl
CAS Number:
Molecular Weight:
166.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

SMILES string

Cl.CC(C)CC(N)C(N)=O

InChI

1S/C6H14N2O.ClH/c1-4(2)3-5(7)6(8)9;/h4-5H,3,7H2,1-2H3,(H2,8,9);1H

InChI key

VSPSRRBIXFUMOU-UHFFFAOYSA-N

Biochem/physiol Actions

DL-Leucinamide hydrochloride is a racemic mixture of D- and L- enantiomers of leucinamide. Leucinamide is an amide derivative of leucine found at the C-terminal of various peptides such as the PAR-2 activating peptide SLIGRL-NH(2).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidenobu Komeda et al.
Applied microbiology and biotechnology, 70(4), 412-421 (2005-07-08)
Brevundimonas diminuta TPU 5720 produces an amidase acting L-stereoselectively on phenylalaninamide. The enzyme (LaaA(Bd)) was purified to electrophoretic homogeneity by ammonium sulfate fractionation and four steps of column chromatography. The final preparation gave a single band on SDS-PAGE with a
Yasuhiko Hashida et al.
Journal of biochemistry, 141(6), 879-888 (2007-04-05)
Thermolysin activity in the hydrolysis of N-[3-(2-furyl)acryloyl]-glycyl-l-leucine amide (FAGLA) and FA-l-leucyl-l-alanine amide (FALAA) was examined at various Co(2+) and Ca(2+) concentrations. It decreased to 28% with increasing [Co(2+)] up to 18 mM. The Co(2+)-dependent inactivation was in part suppressed by
R Pacheco et al.
Analytical biochemistry, 346(1), 49-58 (2005-09-28)
This study demonstrates the use of Fourier transform infrared (FTIR) spectroscopy for monitoring both synthesis and hydrolysis reactions catalyzed by a recombinant amidase (EC 3.5.1.4) from Pseudomonas aeruginosa. The kinetics of hydrolysis of acetamide, propionamide, butyramide, acrylamide, benzamide, phenylalaninamide, alaninamide
Deepak K Bhatt et al.
Headache, 50(6), 1017-1030 (2010-05-22)
The goal of this study was to determine the vascular effects of protease-activated receptor-2 (PAR-2) activation in the rat cranial vasculature. The role of PAR-2 in pain and inflammatory conditions has been established but the information available on its effects
Formation of diastereomeric derivatives of 2-arylpropionic acids using L-leucinamide: lack of generality.
R Mehvar et al.
Journal of chromatography, 431(1), 228-230 (1988-09-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service