Skip to Content
Merck
All Photos(1)

Key Documents

L7783

Sigma-Aldrich

Leu-Arg-Arg-Asp-Leu-Asp-Ala-Ser-Arg-Glu-Ala-Lys-Lys-Gln-Val-Glu-Lys-Ala-Leu-Glu

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C99H175N33O33
CAS Number:
Molecular Weight:
2355.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ~65%

storage temp.

−20°C

SMILES string

CC(C)C[C@H](N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C99H175N33O33/c1-47(2)41-54(103)80(148)118-58(24-18-38-111-97(105)106)85(153)121-60(26-20-40-113-99(109)110)87(155)129-68(45-75(143)144)93(161)128-66(43-49(5)6)92(160)130-67(44-74(141)142)90(158)116-53(11)79(147)131-69(46-133)94(162)122-59(25-19-39-112-98(107)108)86(154)124-62(28-32-71(135)136)82(150)115-51(9)77(145)117-56(22-13-16-36-101)83(151)120-57(23-14-17-37-102)84(152)123-61(27-31-70(104)134)89(157)132-76(50(7)8)95(163)125-63(29-33-72(137)138)88(156)119-55(21-12-15-35-100)81(149)114-52(10)78(146)127-65(42-48(3)4)91(159)126-64(96(164)165)30-34-73(139)140/h47-69,76,133H,12-46,100-103H2,1-11H3,(H2,104,134)(H,114,149)(H,115,150)(H,116,158)(H,117,145)(H,118,148)(H,119,156)(H,120,151)(H,121,153)(H,122,162)(H,123,152)(H,124,154)(H,125,163)(H,126,159)(H,127,146)(H,128,161)(H,129,155)(H,130,160)(H,131,147)(H,132,157)(H,135,136)(H,137,138)(H,139,140)(H,141,142)(H,143,144)(H,164,165)(H4,105,106,111)(H4,107,108,112)(H4,109,110,113)/t51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,76-/m0/s1

InChI key

LDXUYMBPAYIMJR-UGXKBFFBSA-N

Application

Of possible use as an antigen to produce a rheumatic fever vaccine.

Other Notes

Peptide from the conserved region of Group A streptococcal M protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pruksakorn et al.
Lancet (London, England), 344(8923), 639-642 (1994-09-03)
Rheumatic fever and rheumatic heart disease remain very common in developing countries, and a vaccine to protect against these disorders would have a great impact on public health. A vaccine must target the M protein of group A streptococci (Streptococcus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service