Skip to Content
Merck
All Photos(1)

Documents

L6640

Sigma-Aldrich

Lanthanum(III) chloride heptahydrate

ACS reagent, 64.5-70.0% LaCl3 basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3 · 7H2O
CAS Number:
Molecular Weight:
371.37
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

concentration

64.5-70.0% LaCl3

impurities

≤0.01% Insoluble matter

mp

91 °C (dec.) (lit.)

cation traces

Ca: ≤0.001%
Mg: ≤1 ppm

SMILES string

O.O.O.O.O.O.O.Cl[La](Cl)Cl

InChI

1S/3ClH.La.7H2O/h3*1H;;7*1H2/q;;;+3;;;;;;;/p-3

InChI key

FDFPDGIMPRFRJP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Calcium channel blocker

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service