Skip to Content
Merck
All Photos(1)

Key Documents

L5914

Sigma-Aldrich

Laser Tough-Spots®

diam. 3/8 in., yellow

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$255.00

$255.00


Available to ship on07 April 2025Details



Select a Size

Change View
1 PKG
$255.00

About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

$255.00


Available to ship on07 April 2025Details


material

polyolefin
yellow

packaging

pack of 3840 ea (192 labels per sheet)

manufacturer/tradename

Diversified Biotech SPOT-1500

diam.

3/8 in.

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.

Features and Benefits

  • Tough
  • Temperature-resistant
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Fits a wide range of microcentrifuge tube tops, cryovial caps and lids, laboratory container caps, lids, and covers
  • Color-coded for easy sample identification

Legal Information

Tough-Spots is a registered trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service