Skip to Content
Merck
All Photos(1)

Key Documents

L4545

Sigma-Aldrich

L-798106

≥98% (HPLC)

Synonym(s):

(2E)-N-[(5-bromo-2-methoxyphenyl)sulfonyl]-3-[2-(2-naphthalenylmethyl)phenyl]-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$164.00
25 MG
$656.00

$164.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$164.00
25 MG
$656.00

About This Item

Empirical Formula (Hill Notation):
C27H22BrNO4S
CAS Number:
Molecular Weight:
536.44
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

$164.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >20 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

COc1ccc(Br)cc1S(=O)(=O)NC(=O)\C=C\c2ccccc2Cc3ccc4ccccc4c3

InChI

1S/C27H22BrNO4S/c1-33-25-14-13-24(28)18-26(25)34(31,32)29-27(30)15-12-21-7-3-5-9-23(21)17-19-10-11-20-6-2-4-8-22(20)16-19/h2-16,18H,17H2,1H3,(H,29,30)/b15-12+

InChI key

ODTKFNUPVBULRJ-NTCAYCPXSA-N

Application

L-798106, a selective prostanoid receptor EP3 antagonist, is used in prostanoid receptor signaling studies that regulate COX-2 levels and the central excitatory effects of PGE(2) on PVN neurons.[1]

Biochem/physiol Actions

L-798106 is a potent, selective prostanoid receptor EP3-selective antagonists.
L-798106 was among the first prostanoid receptor EP3-selective antagonists. It has been used in multiple studies to tease out EP3 agonist activity, both in vitro and in vivo. It successfully blocks the actions of sulprostone, an EP3-selective agonist, and it helped show that the vascular contraction effect of PGE2 is due to its prostanoid EP3 agonist activity.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexis A Gonzalez et al.
American journal of physiology. Renal physiology, 313(4), F1038-F1049 (2017-07-14)
During the early phase of ANG II-dependent hypertension, tubular PGE
Lixing Zhao et al.
Journal of periodontology, 84(12), 1847-1857 (2013-03-30)
During periodontitis and orthodontic tooth movement, periodontal vasculature is severely impaired, leading to a hypoxic microenvironment of periodontal cells. However, the impact of hypoxia on periodontal cells is poorly defined. The present study investigates responses of cocultured endothelial cells (ECs)
Gen-Lin He et al.
Journal of neuroinflammation, 13(1), 296-296 (2016-11-23)
Prostaglandin E2 (PGE2)-involved neuroinflammatory processes are prevalent in several neurological conditions and diseases. Amyloid burden is correlated with the activation of E-prostanoid (EP) 2 receptors by PGE2 in Alzheimer's disease. We previously demonstrated that electromagnetic field (EMF) exposure can induce
Yiqing Li et al.
Cell death & disease, 11(4), 221-221 (2020-04-07)
T-cell acute lymphoblastic leukaemia (T-ALL) is an aggressive haematological malignancy that is characterized by a high frequency of induction failure and by early relapse. Many studies have revealed that metadherin (MTDH) is highly expressed in a variety of malignant solid
Shoujin Hao et al.
American journal of physiology. Renal physiology, 311(4), F822-F829 (2016-07-29)
The mechanisms by which prostanoids contribute to the maintenance of whole body water homeostasis are complex and not fully understood. The present study demonstrates that an EP3-dependent feedback mechanism contributes to the regulation of water homeostasis under high-salt conditions. Rats

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service