Skip to Content
Merck
All Photos(1)

Key Documents

L2785

Sigma-Aldrich

Lectin from Dolichos biflorus (horse gram)

lyophilized powder

Synonym(s):

Dolichos biflorus agglutinin, DBA

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$116.00
5 MG
$441.00

$116.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
$116.00
5 MG
$441.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$116.00


Estimated to ship on29 May 2025


form

lyophilized powder

potency

≤20 μg per mL agglutination activity

impurities

salt, essentially free

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DBA has anti-A1 human blood group specificity, useful for distinguishing between A1 and A2 blood types. DBA has strong anti-Cad activity, and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.

Preparation Note

Purified by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A1 erythrocytes after 1 hour incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ann Marie Hynes et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9893-9898 (2014-06-21)
Nephronophthisis (NPHP) is the major cause of pediatric renal failure, yet the disease remains poorly understood, partly due to the lack of appropriate animal models. Joubert syndrome (JBTS) is an inherited ciliopathy giving rise to NPHP with cerebellar vermis aplasia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service