Skip to Content
Merck
All Photos(1)

Key Documents

L1415

Sigma-Aldrich

L-162,313

≥95%, solid

Synonym(s):

5,7-Dimethyl-2-ethyl-3-[[4-[2­(n-butyloxycarbonylsulfonamido)-5-isobutyl-3-thienyl]phenyl]methyl]imidazo­[4,5-b]­pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H38N4O4S2
CAS Number:
Molecular Weight:
582.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥95%

form

solid

color

white

solubility

DMSO: soluble 10 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

CCCCOC(=O)NS(=O)(=O)c1sc(CC(C)C)cc1-c2ccc(Cn3c(CC)nc4c(C)cc(C)nc34)cc2

InChI

1S/C30H38N4O4S2/c1-7-9-14-38-30(35)33-40(36,37)29-25(17-24(39-29)15-19(3)4)23-12-10-22(11-13-23)18-34-26(8-2)32-27-20(5)16-21(6)31-28(27)34/h10-13,16-17,19H,7-9,14-15,18H2,1-6H3,(H,33,35)

InChI key

RINPELQWLUGERM-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)
rat ... Agtr1(24180)

Biochem/physiol Actions

Non-peptide AT1 receptor agonist.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service