Skip to Content
Merck
All Photos(2)

Key Documents

L133

Sigma-Aldrich

L-750,667 trihydrochloride

solid

Synonym(s):

3-[4-(4-Iodophenyl)piperazin-1-yl]methyl-1H-pyrrolo[2,3-b]pyridine trihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19IN4 · 3HCl
CAS Number:
Molecular Weight:
527.66
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

DMSO: 10 mg/mL

SMILES string

Cl.Cl.Cl.Ic1ccc(cc1)N2CCN(CC2)Cc3c[nH]c4ncccc34

InChI

1S/C18H19IN4.3ClH/c19-15-3-5-16(6-4-15)23-10-8-22(9-11-23)13-14-12-21-18-17(14)2-1-7-20-18;;;/h1-7,12H,8-11,13H2,(H,20,21);3*1H

InChI key

YOURSPNOEWYAKO-UHFFFAOYSA-N

Gene Information

human ... DRD4(1815)

Application

Useful in characterizing the extracellular domain of the D4 dopamine receptor.[1]

Biochem/physiol Actions

Selective D4 dopamine receptor antagonist.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Schetz et al.
Molecular pharmacology, 57(1), 144-152 (2000-01-05)
The molecular determinants that govern selective ligand binding to the rat D(4) dopamine receptor were investigated by substituting D(2) dopamine receptor sequences into a D(4) dopamine receptor background. The resulting mutant D(4) dopamine receptors were then screened with a panel
Andrew M Bailey et al.
Journal of bacteriology, 192(6), 1607-1616 (2010-01-19)
The transcriptomes of Salmonella enterica serovar Typhimurium SL1344 lacking a functional ramA or ramR or with plasmid-mediated high-level overexpression of ramA were compared to those of the wild-type parental strain. Inactivation of ramA led to increased expression of 14 SPI-1
S Patel et al.
Molecular pharmacology, 50(6), 1658-1664 (1996-12-01)
We identified a novel azaindole derivative, L-750,667, that has high affinity (Ki = 0.51 nM) and >2000-fold selectivity for D4 dopamine receptors compared with its activity at D2 and D3 dopamine receptors. L-750,667 had little affinity for rat D1/D5 dopamine
Margareta K Sorenson et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(45), 16722-16727 (2006-11-01)
The dissociable sigma subunit of bacterial RNA polymerase is required for the promoter-specific initiation of transcription. When bound to RNA polymerase, sigma makes sequence-specific promoter contacts and plays a crucial role in DNA melting. In isolation, however, sigma lacks significant
Takashi Iwase et al.
Circulation, 111(3), 356-362 (2005-01-19)
Previous studies have shown that adrenomedullin (AM) inhibits vascular endothelial cell apoptosis and induces angiogenesis. We investigated whether AM enhances bone marrow cell-induced angiogenesis. Immediately after hindlimb ischemia was created, rats were randomized to receive AM infusion plus bone marrow-derived

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service