Skip to Content
Merck
All Photos(2)

Key Documents

L0633

Sigma-Aldrich

Luciferase-luciferin

suitable for determination of ATP released from blood platelets., powder

Synonym(s):

Luciferase-Luciferin reagent preparation from firefly

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200

form

powder

usage

100 mg sufficient for 50 assays (with cell extract or platelets)

ATP measuring range

10-15 mole (using 100 μl of reagent (20 mg/ml))

suitability

suitable for determination of ATP released from blood platelets.

storage temp.

−20°C

Biochem/physiol Actions

A complete reagent designed for the determination of ATP generated in coupled enzyme systems or present in cell extracts.

Other Notes

Contains luciferase, luciferin, MgSO4, EDTA and buffer salts in a human albumin base.

Physical form

in glycine buffer

Reconstitution

Reconstitute with sterile water.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service