Skip to Content
Merck
All Photos(1)

Documents

K4268

Sigma-Aldrich

PKCμ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PKCM, PKD, PRKCM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

578-782 nmol/min·mg

mol wt

~131 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKD1(5587)

General description

Protein kinase C μ (PKCμ) belongs to the protein kinase C (PKC) family. PKCμ is widely expressed with the highest expression in the thymus, lung and peripheral blood mononuclear cells.

Biochem/physiol Actions

Protein kinase C μ (PKCμ) associates with phosphatidylinositol 4-kinase as well as phosphatidylinositol-4-phosphate 5-kinase. A region of PKCμ between the amino-terminal transmembrane domain and the pleckstrin homology domain is shown to be involved in the association with the lipid kinases.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Nishikawa et al.
The Journal of biological chemistry, 273(36), 23126-23133 (1998-08-29)
Protein kinase Cmu (PKCmu), also named protein kinase D, is an unusual member of the PKC family that has a putative transmembrane domain and pleckstrin homology domain. This enzyme has a substrate specificity distinct from other PKC isoforms (Nishikawa, K.
J Rennecke et al.
European journal of biochemistry, 242(2), 428-432 (1996-12-01)
13 murine tissues and 12 cell lines were tested for the expression of the novel protein kinase C (PKC) isoenzyme mu. Using two different PKC mu antibodies (sc-639 and P26720), PKC mu was detected in all tissues and cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service