Skip to Content
Merck
All Photos(1)

Key Documents

K4143

Sigma-Aldrich

PKCη, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MG C26269, MG C5363, PKCL, PRK CL, PRKCH, nPKC-eta

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$419.00

$419.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$419.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$419.00


Estimated to ship on21 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

73-99 nmol/min·mg

mol wt

~73 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCH(5583)

General description

PKCη (protein kinase C η) belongs to the protein kiinase C family. It is mainly expressed in the squamous cell epithelia.[1]

Biochem/physiol Actions

PKCη (protein kinase C η) mainly participates in terminal differentiation of keratinocytes. It is responsible for arresting the cell cycle at G1 phase. It is also involved in phosphorylation of p21 and suppression of cdk2 (cyclin-dependent kinase 2) kinase activity by associating with cyclin E/cdk2/p21 complex, thereby regulating cell cycle.[1] PKCη is upregulated in breast cancer and is associated with cell growth.[2]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kashiwagi et al.
Oncogene, 19(54), 6334-6341 (2001-02-15)
PKC is activated on the cell membrane by phospholipids, thereby transducing signals to intracellular pathways. We provide here another function of PKC, namely, regulating cell cycle by interaction with the cyclin E/cdk2/p21 complex. Among the 10 isoforms of PKC, PKCeta
Deepanwita Pal et al.
Biochimica et biophysica acta, 1830(8), 4040-4045 (2013-04-09)
Protein kinase C (PKC) serves as the receptor for tumor-promoting phorbol esters, which are potent activators of conventional (c) and novel (n) PKCs. We recently showed that these activators induced selective upregulation of PKCη in breast cancer cells. The objective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service