Skip to Content
Merck
All Photos(1)

Key Documents

K4019

Sigma-Aldrich

KHS101

≥98% (HPLC)

Synonym(s):

N4-isobutyl-N2-((2-phenylthiazol-4-yl)methyl)pyrimidine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5S
CAS Number:
Molecular Weight:
339.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)CNC1=NC(NCC2=CSC(C3=CC=CC=C3)=N2)=NC=C1

InChI

1S/C18H21N5S/c1-13(2)10-20-16-8-9-19-18(23-16)21-11-15-12-24-17(22-15)14-6-4-3-5-7-14/h3-9,12-13H,10-11H2,1-2H3,(H2,19,20,21,23)

InChI key

DGRJOOOHPBSAHD-UHFFFAOYSA-N

Biochem/physiol Actions

KHS101 is a small molecule differentiator of neuronal differentiation: TACC3 binder.
KHS101 is a small molecule that drives neuronal differentiation of hippocampal neuronal progenitor cells (NPC). Treatment of NPCs with KHS101 induces expression of the neurogenic transcription factor NeuroD and the neuronal marker Tuj1, and exhibit neuronal morphology and spiking activity (EC50 1 uM). KHS101 binds tightly to TACC3, which is a structural component of the centrosome and mitotic spindle apparatus. TACC3 loss of function studies in mice and stem cells suggest that TACC3 controls progenitor cell expansion and terminal differentiation during development.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Petschnigg et al.
Journal of molecular biology, 429(2), 280-294 (2016-12-14)
The epidermal growth factor receptor (EGFR) is a receptor tyrosine kinase (TK) that-once activated upon ligand binding-leads to receptor dimerization, recruitment of protein complexes, and activation of multiple signaling cascades. The EGFR is frequently overexpressed or mutated in various cancers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service