Skip to Content
Merck
All Photos(1)

Key Documents

K2768

Sigma-Aldrich

KHS1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GCKR, MAP4K5, MAPKKKK5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

76-99 nmol/min·mg

mol wt

~135 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP4K5(11183)

Biochem/physiol Actions

KHS1 (kinase homologous to SPS1/STE20) is a component of MAP (mitogen activated protein) kinase signaling pathway and is responsible for activation of JUNK (c-Jun N-terminal kinase). Mutation in the promoter region of KHS1 is linked with lower risk of type 2 diabetes. It is also referred to as MAP4K5 (mitogen-activated protein kinase kinase kinase kinase 5)

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maura Epifanía Matus-Ortega et al.
Biochimica et biophysica acta, 1769(3), 194-203 (2007-03-06)
PNPase is a phosphate-dependent exonuclease of Escherichia coli required for growth in the cold. In this work we explored the effect of specific mutations in its two RNA binding domains KH and S1 on RNA binding, enzymatic activities, autoregulation and
C S Shi et al.
Journal of immunology (Baltimore, Md. : 1950), 163(6), 3279-3285 (1999-09-08)
TNF-induced activation of stress activated protein kinases (SAPKs, Jun NH2-terminal kinases) requires TNF receptor associated factor 2 (TRAF2). TRAF2 is a potent activator of a 95-kDa serine/threonine kinase termed germinal center kinase related (GCKR, also referred to as KHS1), which
Yanyun Gu et al.
Journal of human genetics, 51(7), 605-610 (2006-05-16)
MAP4K5 (mitogen-activated protein kinase kinase kinase kinase 5), an early component of MAP kinase signal cascades was shown to activate Jun kinase in mammalian cells. The association between SNPs of MAP4K5 and type 2 diabetes (T2DM) was investigated due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service