Skip to Content
Merck
All Photos(1)

Key Documents

I9141

Sigma-Aldrich

Anti-Interferon-γ antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IFN-γ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ifng(15978)

Looking for similar products? Visit Product Comparison Guide

General description

Interferons (IFNs) are cytokines that are secreted in response to viral or bacterial infections and tumorigenesis. There are two types of IFNs, type I (IFN-α and IFN-β) and type II (IFN-γ). IFN-γ binds to specific receptor complex consisting of IFNγR1 and IFNγR2. There are many pathways that are mediated by IFN-γ binding such as JAK/STAT1, AP-1, NF-κB, STAT3 and STAT5. IFN-γ is pleotropic and performs various functions related to immune response, inflammation, differentiation and activation of T cells, cell cycle and apoptosis. The most studied function of IFN-γ is priming the antigen presenting cells by upregulating major histocompatibility (MHC) Class I molecules. IFN-γ has clinical applications in autoimmune diseases (rheumatoid arthritis), multiple sclerosis, cancer, HIV and fungal infections. Anti-Interferon-γ recognizes mouse Interferon-γ. It shows less than 1% cross reactivity with human IFN-γ.

Specificity

The antibody shows <1% cross-reactivity with recombinant human IFN-γ.

Immunogen

recombinant mouse IFN-γ.

Application

Anti-interferon-γ antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. The antibody is suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline (pH 7.4) with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Sekine et al.
Cell, 183(1), 158-168 (2020-09-28)
SARS-CoV-2-specific memory T cells will likely prove critical for long-term immune protection against COVID-19. Here, we systematically mapped the functional and phenotypic landscape of SARS-CoV-2-specific T cell responses in unexposed individuals, exposed family members, and individuals with acute or convalescent COVID-19. Acute-phase
Adithya Cattamanchi et al.
Journal of acquired immune deficiency syndromes (1999), 56(3), 230-238 (2011-01-18)
To determine whether interferon-gamma release assays (IGRAs) improve the identification of HIV-infected individuals who could benefit from latent tuberculosis infection therapy. Systematic review and meta-analysis. We searched multiple databases through May 2010 for studies evaluating the performance of the newest
Natalia Lubina-Dąbrowska et al.
Archivum immunologiae et therapiae experimentalis, 65(4), 325-338 (2017-03-17)
The aim of this study was to investigate the effects of interferon (IFN)-β1a and IFN-β1b treatment on inflammatory factors and myelin protein levels in the brain cortex of the Lewis rat experimental autoimmune encephalomyelitis (EAE), animal model of multiple sclerosis.
Leonidas C Platanias
Nature reviews. Immunology, 5(5), 375-386 (2005-05-03)
Interferons are cytokines that have antiviral, antiproliferative and immunomodulatory effects. Because of these important properties, in the past two decades, major research efforts have been undertaken to understand the signalling mechanisms through which these cytokines induce their effects. Since the
S E Ealick et al.
Science (New York, N.Y.), 252(5006), 698-702 (1991-05-03)
The x-ray crystal structure of recombinant human interferon-gamma has been determined with the use of multiple-isomorphous-replacement techniques. Interferon-gamma, which is dimeric in solution, crystallizes with two dimers related by a noncrystallographic twofold axis in the asymmetric unit. The protein is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service