I7904
Indomethacin heptyl ester
methyl acetate solution, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H30ClNO4
CAS Number:
Molecular Weight:
455.97
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98% (HPLC)
form
methyl acetate solution
potency
0.04 μM IC50 (for human recombinat COX-2)
shipped in
wet ice
storage temp.
−20°C
SMILES string
CCCCCCCOC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13
InChI
1S/C26H30ClNO4/c1-4-5-6-7-8-15-32-25(29)17-22-18(2)28(24-14-13-21(31-3)16-23(22)24)26(30)19-9-11-20(27)12-10-19/h9-14,16H,4-8,15,17H2,1-3H3
InChI key
PYBCHCVNKGZCOH-UHFFFAOYSA-N
Application
Used to inhibit COX-1 and COX-2. To achieve 50% inhibition of human recombinant COX-2 use I7904 at 0.04 uM/mL .
Biochem/physiol Actions
Potent cyclooxygenase-2 inhibitor. 1700 times greater activity against COX-2 than COX-1.
Other Notes
Derivatized indomethacin.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
8.6 °F - closed cup
Flash Point(C)
-13 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service