Skip to Content
Merck
All Photos(1)

Documents

I6904

Sigma-Aldrich

Anti-Inositol Hexakisphosphate Kinase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IP6 Kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

mouse, rabbit, rat, human

technique(s)

western blot: suitable

shipped in

wet ice

storage temp.

−20°C

General description

The gene IP6K1 (inositol hexakisphosphate kinase 1), also referred to as IHPK1, is mapped to human chromosome 3p21.31. The 1323bp open reading frame contains six exons that encode a protein of 441 amino acids. It is found to be expressed in several tissues, such as testis, brain, heart, pancreas, liver, kidney, and skeletal muscle.

Immunogen

synthetic peptide corresponding to amino acids 216-233 of human IP6 kinase.

Biochem/physiol Actions

The gene IP6K1 (inositol hexakisphosphate kinase 1) encodes a member of the inositol hexakisphosphate kinases (InsP6K) that also comprise of InsP6K2, and InsP6K3. The members of this family are involved in the conversion of InsP6 (phytic acid) to InsP7 (diphosphoinositol-pentakisphosphate), which functions in vesicular trafficking and exocytosis, apoptosis, and insulin disposition. Knock-out of this gene in mice has been shown to increase insulin sensitivity due to a decrease in the levels of InsP7, a physiologic inhibitor of Akt, a serine/threonine protein kinase that regulates glucose homeostasis. IP6K1 is found to negatively regulate PtdIns(3,4,5)P3 signaling, which in turn, regulates the function of neutrophils in innate immunity.

Physical form

100 μg of affinity isolated IgG in PBS with 1.0 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junichi Kamimura et al.
Journal of human genetics, 49(7), 360-365 (2004-06-29)
Type 2 diabetes mellitus (T2DM) is a group of multifactorial disorders due to either defective insulin secretion or action. Despite the fact that numerous genetic researches of T2DM have been pursued, the pathogenic mechanisms remain obscure. We encountered a T2DM
Amit Prasad et al.
Nature immunology, 12(8), 752-760 (2011-06-21)
Inositol phosphates are widely produced throughout animal and plant tissues. Diphosphoinositol pentakisphosphate (InsP7) contains an energetic pyrophosphate bond. Here we demonstrate that disruption of inositol hexakisphosphate kinase 1 (InsP6K1), one of the three mammalian inositol hexakisphosphate kinases (InsP6Ks) that convert

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service