Skip to Content
Merck
All Photos(1)

Key Documents

I3901

Sigma-Aldrich

IL-1α rat

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-1α, Lymphocyte activating factor, rIL-1α, IL-1α

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$879.00

$879.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$879.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$879.00


Estimated to ship on12 April 2025


Request a Bulk Order

Product Name

Interleukin-1α from rat, IL-1α, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1-10 pg/mL ED50

quality

endotoxin tested

mol wt

~18 kDa

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤0.1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Il1a(24493)

Biochem/physiol Actions

Interleukin-1 (IL-1), originally known as Lymphocyte Activating Factor (LA F), activates T cells and lymphocytes, which then proliferate and secrete Interleukin-2 (IL-2). The proliferative activity of human IL-1α is tested in culture using the murine cell line D10S.

Physical form

Lyophilized from 0.2 μm filtered phosphate buffered saline containing 0.25 mg of bovine serum albumin.

Analysis Note

The biological activity of recombinant rat IL-1α was measured by its ability to stimulate cell proliferation using a mouse helper T cell line, D10.G4.1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuankang Wang et al.
Frontiers in neuroscience, 15, 768262-768262 (2021-11-20)
After spinal cord injury (SCI), reactive astrocytes can be classified into two distinctive phenotypes according to their different functions: neurotoxic (A1) astrocytes and neuroprotective (A2) astrocytes. Our previous studies proved that photobiomodulation (PBM) can promote motor function recovery and improve
Xuankang Wang et al.
Journal of neuroinflammation, 18(1), 256-256 (2021-11-07)
Neurotoxic microglia and astrocytes begin to activate and participate in pathological processes after spinal cord injury (SCI), subsequently causing severe secondary damage and affecting tissue repair. We have previously reported that photobiomodulation (PBM) can promote functional recovery by reducing neuroinflammation
T Nishida et al.
Journal of biochemistry, 105(3), 351-357 (1989-03-01)
A cDNA sequence coding for rat interleukin-1 alpha (IL-1 alpha) has been isolated from a cDNA library that was prepared with mRNA derived from LPS-stimulated rat peritoneal macrophages by using human IL-1 alpha cDNA as a probe. The rat cDNA
Sualyneth Galarza et al.
Advanced healthcare materials, 9(4), e1901419-e1901419 (2020-01-17)
Bioengineers have designed numerous instructive brain extracellular matrix (ECM) environments with tailored and tunable protein compositions and biomechanical properties in vitro to study astrocyte reactivity during trauma and inflammation. However, a major limitation of both protein-based and synthetic model microenvironments
Shuqi Jiang et al.
Journal of inflammation research, 14, 5919-5937 (2021-11-23)
Astrocyte A1/A2 phenotypes may play differential role in the pathogenesis of periventricular white matter (PWM) damage in septic postnatal rats. This study aimed to determine whether melatonin (MEL) would improve the axonal hypomyelination through shifting A1 astrocytes towards A2. One-day-old

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service