Skip to Content
Merck
All Photos(1)

Key Documents

I3659

Sigma-Aldrich

Monoclonal Anti-ING1 antibody produced in mouse

~2 mg/mL, clone Cab 3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Inhibitor of Growth 1, Anti-p33ING1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Cab 3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~33 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 2-4 μg/mL using K562 total cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ING1(3621)

General description

Three alternatively spliced mRNAs of the gene ING1 (inhibitor of growth family member 1), p47ING1a, p33ING1b and p24ING1c, encode for proteins that function as growth inhibitors. The proteins are localized to the nucleus and they have a common C-terminal region. The major isoforms expressed in humans are p47ING1a and p33ING1b. The gene ING1 is mapped to human chromosome 13q34.

Immunogen

recombinant ING1.

Biochem/physiol Actions

The ING (inhibitor of growth) family of proteins is involved in several cellular processes, such as cell cycle, senescence and apoptosis. They activate acetylation-dependent pathways that in turn, regulate gene expression. The isoform p33ING1b is found to be involved in the regulation of apoptosis. Suppression of p33ING1b expression is found to promote the formation of tumor. On the other hand, increased expression arrests cells at G1 phase of the cell cycle and inhibits the progression of cell cycle. These processes are mediated by interactions with p53 tumor suppressor. Human ING1 proteins are found to be associated with proteins involved in histone acetyltransferase (HAT) activity, such as TRRAP (transformation/transcription domain associated protein), PCAF (P300/CBP-associated factor), CBP (cap binding protein), and p300. The isoform p47ING1a is found to inhibit histone acetylation and bind to histone deacetylase HDAC1 (histone deacetylase 1). The isoform p33ING1b regulates the association between proliferating cell nuclear antigen (PCNA) and p30 that is important for DNA repair and chromatin remodeling.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Boland et al.
Hybridoma, 19(2), 161-165 (2000-06-27)
Nine monoclonal antibodies (MAbs) against human and rodent ING1 protein have been generated using an IL6-secreting mouse myeloma line. These antibodies are all effective in recognizing ING1 protein in ELISAs, Western blot assays, and by indirect immunofluorescence. Combining different CAb
Diego Vieyra et al.
Cancer research, 62(15), 4445-4452 (2002-08-03)
Recently, several novel human ING1 isoforms have been cloned. However,the biochemical functions and the involvement of these proteins in apoptosis remain uncharacterized. We have examined the apoptotic effects and biochemical functions of the two major human ING1 isoforms p47(ING1a) and
M Zeremski et al.
Somatic cell and molecular genetics, 23(3), 233-236 (1997-05-01)
A novel gene ING1 was recently cloned and defined as a candidate tumor suppressor gene. Reduced expression and rearrangements of ING1 are found in several tumor cell lines, ING1 overexpression is associated with cell growth arrest and ING1 suppression promotes
Hiromi Kataoka et al.
Cancer research, 63(18), 5785-5792 (2003-10-03)
The ING family of proteins is involved in the regulation of diverse processes ranging from cell cycle and cellular senescence to apoptosis. These effects are most likely through activation of acetylation-dependent pathways that ultimately alter gene expression. Despite reports linking
M Nagashima et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(17), 9671-9676 (2001-08-02)
The p33ING1 protein is a regulator of cell cycle, senescence, and apoptosis. Three alternatively spliced transcripts of p33ING1 encode p47ING1a, p33ING1b, and p24ING1c. We cloned an additional ING family member, p33ING2/ING1L. Unlike p33ING1b, p33ING2 is induced by the DNA-damaging agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service