Skip to Content
Merck
All Photos(1)

Key Documents

I131

Sigma-Aldrich

D-myo-Inositol 1,3,4,5-tetrakis(phosphate) potassium salt

Synonym(s):

IP4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$414.00

$414.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$414.00

About This Item

Empirical Formula (Hill Notation):
C6H8O18P4K8
CAS Number:
Molecular Weight:
804.80
UNSPSC Code:
12352302
NACRES:
NA.25
grade:
for analytical purposes

$414.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

for analytical purposes

storage temp.

−20°C

SMILES string

[K+].[K+].[K+].[K+].[K+].[K+].[K+].[K+].[P](=O)([O-])([O-])O[C@@H]1[C@H](C([C@@H]([C@@H](C1O[P](=O)([O-])[O-])O[P](=O)([O-])[O-])O)O[P](=O)([O-])[O-])O

InChI

1S/C6H16O18P4.8K/c7-1-3(21-25(9,10)11)2(8)5(23-27(15,16)17)6(24-28(18,19)20)4(1)22-26(12,13)14;;;;;;;;/h1-8H,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20);;;;;;;;/q;8*+1/p-8/t1-,2-,3?,4-,5+,6?;;;;;;;;/m0......../s1

InChI key

HDLWTQSZUMYYND-QAVKKEKOSA-F

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Precursor of inositol 1,3,4-trisphosphate; increases entry of calcium across the plasma membrane; proposed regulator of intracellular calcium by control of plasma membrane transport of calcium ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serena Casanova et al.
ACS applied materials & interfaces, 11(1), 1689-1698 (2018-12-14)
The independent effect of nanotube surface chemistry and structure on the flow of water under nanoscale confinement is demonstrated in this paper for the first time via the synthesis of novel carbon nitride nanotube (CNNT) membranes. Using a combination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service