Skip to Content
Merck
All Photos(1)

Key Documents

I0144

Sigma-Aldrich

Indolicidin

≥97% (HPLC)

Synonym(s):

Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C100H132N26O13
CAS Number:
Molecular Weight:
1906.28
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N6CCC[C@H]6C(=O)N[C@@H](Cc7c[nH]c8ccccc78)C(=O)N[C@@H](Cc9c[nH]c%10ccccc9%10)C(=O)N%11CCC[C@H]%11C(=O)N[C@@H](Cc%12c[nH]c%13ccccc%12%13)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

General description

Chemical structure: peptide

Biochem/physiol Actions

Exhibits potent antimicrobial activity in vitro against bacteria and fungi.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Kovacs-Nolan et al.
The Journal of general virology, 90(Pt 8), 1892-1905 (2009-04-24)
Respiratory syncytial virus (RSV) is the leading cause of serious respiratory tract disease in children and calves; however, RSV vaccine development has been slow due to early observations that formalin-inactivated vaccines induced Th2-type immune responses and led to disease enhancement
Tatyana I Rokitskaya et al.
Biochimica et biophysica acta, 1808(1), 91-97 (2010-09-21)
Indolicidin, a 13-residue cationic peptide with extremely high tryptophan content, exhibits broad-spectrum antimicrobial as well as hemolytic activity. To gain insight into the mechanism of indolicidin action on membrane permeability, liposome leakage induced by this peptide was studied by using
J Kovacs-Nolan et al.
Vaccine, 27(14), 2055-2064 (2009-05-12)
The need to enhance the immunogenicity of purified subunit antigens and modulate resulting immune responses has prompted the development of new adjuvants. Here, the ability of CpG oligodeoxynucleotides (ODN), a bovine host defence peptide indolicidin, and polyphosphazene to synergistically combine
Setsuko Ando et al.
Journal of peptide science : an official publication of the European Peptide Society, 16(4), 171-177 (2010-03-03)
A series of Trp and Arg analogs of antibacterial indolicidin (Ind) was synthesized and the antimicrobial and hemolytic activities were investigated. [L(9)]Ind, [L(11)]Ind, [K(8),L(9)]Ind and [K(6, 8),L(9)]Ind showed desirable characteristics, exhibiting negligible hemolytic activity while keeping strong antibacterial activity. The
Linda Tomasinsig et al.
Infection and immunity, 78(4), 1781-1788 (2010-01-27)
Cathelicidins are peptide components of the innate immune system of mammals. Apart from exerting a direct antibiotic activity, they can also trigger specific defense responses in the host. Their roles in various pathophysiological conditions have been studied, but there is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service