Key Documents
HS1421A
Weigh Boat
Small, square blue polystyrene, pk of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Select a Size
500 EA
$69.70
$69.70
Available to ship on14 April 2025Details
Select a Size
Change View
500 EA
$69.70
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
$69.70
Available to ship on14 April 2025Details
material
square blue polystyrene
size
small
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1421A
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer
Choose from one of the most recent versions:
Certificates of Analysis (COA)