Skip to Content
Merck
All Photos(1)

Key Documents

HS120732

Reagent Reservoir Chiller

capacity 50 mL, purple/blue

Sign Into View Organizational & Contract Pricing

Select a Size

2 EACH
$134.00

$134.00


Check Cart for Availability


Select a Size

Change View
2 EACH
$134.00

About This Item

UNSPSC Code:
41121522
NACRES:
NB.04

$134.00


Check Cart for Availability

material

polypropylene
purple/blue

feature

to hold and chill reagent reservoirs, basins

packaging

pkg of 2 ea

manufacturer/tradename

Heathrow Scientific 120732

L × W × H

144 mm × 74 mm × 45 mm

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

The Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.

Features and Benefits

  • Reagents are kept at less than 5°C (41°F) for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Stackable, save space when storing in the freezer or on the bench top
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Made from sonic welded, engineered polypropylene that ensures a leak free design
  • Not intended for autoclave, do not use with liquid nitrogen
  • Reusable
  • Two-year warranty

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service