Skip to Content
Merck
All Photos(2)

Key Documents

HS120372

TrueNorth® Flatpack PP Thin Film Freezer Boxes

Holds 81 x 1.5-2 mL tubes, green, pk of 10

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$152.00

$152.00


Available to ship on22 April 2025Details



Select a Size

Change View
10 EA
$152.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.22

$152.00


Available to ship on22 April 2025Details


material

polypropylene

feature

Holds 81 x 1.5-2 mL tubes

packaging

pk of 10

manufacturer/tradename

Heathrow Scientific HS120372

technique(s)

cryopreservation: suitable

well diam.

12.1 mm

color

green

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • made from polypropylene material
  • easy self-assemble boxes shipped flat-packed
  • more durable than cardboard boxes as they are not susceptible to moisture
  • significantly reduces the change of mold
  • dividers included with the boxes
  • store down to temperatures of -80C

Legal Information

TrueNorth is a registered trademark of Heathrow Scientific LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service