Skip to Content
Merck
All Photos(2)

Key Documents

HS120318

Vortex Mixer

AC/DC input 100 V / 120 V, Japan plug, blue

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$501.00

$501.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$501.00

About This Item

UNSPSC Code:
41103814

$501.00


Check Cart for Availability

material

blue

packaging

pkg of 1 ea

manufacturer/tradename

Heathrow Scientific 120318

Looking for similar products? Visit Product Comparison Guide

General description

Multi-purpose, maximum convenience

One mixing head does it all with this sleek, modern and compact design vortex mixer!

Mixing Head Holding Capacity:
  • Center pad area for hand held mixing
  • Assay plate x 1 (to be used with retention cords)
  • 50 mL conical tubes x 2 (horizontally mounted)
  • 15 mL conical tubes x 2 (horizontally mounted)
  • 5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL)
  • 1.5/2 mL microfuge tubes x 4 (vertically mounted)
  • 0.5 mL microfuge tubes x 6 (vertically mounted)
  • 0.2 mL microfuge tubes (strip tubes) x 24 (vertical mounted)


Control switch with three operation selections (intermittent, off, continuous).
Infinite position rotary speed control from Off up to approx 3000 rpm

Made of durable, easy to clean ABS material and stable elastomer feet stabilize the unit and dampen vibration.

Packaging

INCLUDES:
1 Mini Vortexer
2 Silicone Caps

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service