Skip to Content
Merck
All Photos(2)

Key Documents

HS120078

DuraPorter® specimen transport box

yellow

Synonym(s):

DuraPorter Transport Box

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$181.00

$181.00


Available to ship on15 April 2025Details



Select a Size

Change View
1 EACH
$181.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.17

$181.00


Available to ship on15 April 2025Details


material

polycarbonate

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS120078

size

380x196x160 mm

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

The DuraPorter Specimen or Sample Transport Box is ideal for transporting biological and clinical samples, as well as instruments and products that you want to keep clean and dry under testing environmental conditions. The clarity of the DuraPorter(R) enables the user to visibly check sample integrity, so that it can be opened using the right precautions. Polycarbonate construction and avaiable in 3 colors for user/ ownership identification. Silicone water-tight seal and three lid clasps provide a secure closure. Carry handle and folds neatly into body space allowing the units to be stacked. Lid opens a full 180 ° allowing for total access to contents and making it easier to clean. Will accomodate 13 and 16mm tubes in a 72-place unwire rack or similar sized rack.
Unit is autoclavable.

Legal Information

Duraporter is a registered trademark of Heathrow Scientific

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service