Skip to Content
Merck
All Photos(4)

Key Documents

HPA055619

Sigma-Aldrich

Anti-DEPDC5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

DEPDC5 Antibody - Anti-DEPDC5 antibody produced in rabbit, Depdc5 Antibody, Anti-DEP domain containing 5, Anti-DEP.5, Anti-KIAA0645

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

YDAQVFRLPGPSRAQCLTTCRSVRERESHSRKSASSCDVSSSPSLPSRTLPTEEVRSQASDDSSLGKSANILMIPHPHLHQYEVSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DEPDC5(9681)

General description

The DEP domain containing 5 (DEPDC5) gene encodes DEP domain-containing protein 5. DEPDC5 protein is made of five functional domains, such as N-terminal domain, a steric hindrance for enhancement of nucleotidase activity domain, disheveled, egl-10, and pleckstrin (DEP) domain, the structural axis for binding arrangement domain, and C-terminal domain. The DEPDC5 gene is located on the human chromosome at 22q12.2-q12.3.

Immunogen

DEP domain containing 5 recombinant protein epitope signature tag (PrEST)

Application

Anti-DEPDC5 antibody produced in rabbit has been used in immunofluorescence (1:100).[1]

Biochem/physiol Actions

DEP domain containing 5 (DEPDC5) acts as a negative regulator of mammalian target of rapamycin complex 1 (mTORC1). It is a part of the GTPase-activating protein activity. Mutations in the DEPDC gene that lead to autosomal dominant familial focal epilepsy with variable foci (FFEVF), familial temporal lobe epilepsy, and autosomal dominant nocturnal frontal lobe epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78172

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Liu et al.
Frontiers in neuroscience, 14, 821-821 (2020-08-28)
To explore the phenotype spectrum of DEPDC5 variants and the possible mechanisms underlying phenotypical variation, we performed targeted next-generation sequencing in 305 patients with focal epilepsies and 91 patients with generalized epilepsies. Protein modeling was performed to predict the effects
Stéphanie Baulac et al.
Annals of neurology, 77(4), 675-683 (2015-01-28)
The DEPDC5 (DEP domain-containing protein 5) gene, encoding a repressor of the mTORC1 signaling pathway, has recently emerged as a major gene mutated in familial focal epilepsies. We aimed to further extend the role of DEPDC5 to focal cortical dysplasias
Ya-Chin Wang et al.
Hepatology communications, 6(12), 3563-3577 (2022-11-03)
Hepatocellular carcinoma (HCC) is a highly invasive malignancy. Recently, GATOR1 (Gap Activity TOward Rags 1) complexes have been shown to play an important role in regulating tumor growth. NPRL2 is a critical component of the GATOR1 complex. Therefore, this study
Fábio A Nascimento et al.
Neurology. Genetics, 1(4), e28-e28 (2016-04-12)
The DEPDC5 gene (OMIM #614191), mapped to 22q12.2-q12.3, encodes the DEP domain-containing protein 5. DEPDC5 has been associated with a variety of familial epilepsies, including familial focal epilepsy with variable foci, autosomal dominant nocturnal frontal lobe epilepsy, familial temporal lobe
Yuki Mizuno et al.
Scientific reports, 8(1), 106-106 (2018-01-10)
Decrease in blood concentration of branched-chain amino acids, especially leucine, is known to promote liver carcinogenesis in patients with chronic liver disease, but the mechanism is unclear. We herein established hepatocellular carcinoma (HCC) cells knocked out for DEPDC5 by using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service