Skip to Content
Merck
All Photos(5)

Key Documents

HPA052026

Sigma-Aldrich

Anti-PNLDC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ40240, Anti-dJ195P10.2, Anti-poly(A)-specific ribonuclease (PARN)-like domain containing 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VGHNMMMDLLHLHEKFFRPLPESYDQFKQNIHSLFPVLIDTKSVTKDIWKEMNFPRVSNLSEVYEVLNSDLNPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNLDC1(154197)

General description

The gene PNLDC1 (PARN like, ribonuclease domain containing 1) is mapped to human chromosome 6q. It is a homologue of PARN (poly(A)-specific ribonuclease). The protein is present in the cytoplasm of stem and spermatogenic cells. The protein has a putative transmembrane domain. The PNLDC1 protein also contains an RNaseH-like domain and RNAse_CAF1 (CCR4-associated factor 1) domain. PNLDC1 is mainly expressed in the early stages of development and upon differentiation.[1]

Immunogen

poly(A)-specific ribonuclease (PARN)-like domain containing 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PNLDC1 (PARN like, ribonuclease domain containing 1) is a Mg2+-dependent exonuclease. In silkworm (Bombyx mori), it is responsible for 3′ pre-piRNA trimming. In humans, PNLDC1 exhibits 3′-5′ exonuclease activity under in vitro conditions. This activity is specific for DNA or RNA polyadenylates. PNLDC1 might be involved in the maintenance of genome integrity and posttranscriptional regulation and checking.[1] It might also be associated with affective psychosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85571

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A balanced translocation disrupting BCL2L10 and PNLDC1 segregates with affective psychosis.
Bouwkamp CG, et al.
American Journal of Medical Genetics. Part B, Neuropsychiatric Genetics : the Official Publication of the International Society of Psychiatric Genetics, doi: 10-doi: 10 (2016)
Differential patterns of allelic loss in estrogen receptor-positive infiltrating lobular and ductal breast cancer.
Loo LW, et al.
Genes Chromosomes Cancer, 47, 1049-1066 (2008)
Dimitrios Anastasakis et al.
Nucleic acids research, 44(18), 8908-8920 (2016-08-16)
PNLDC1 is a homologue of poly(A) specific ribonuclease (PARN), a known deadenylase with additional role in processing of non-coding RNAs. Both enzymes were reported recently to participate in piRNA biogenesis in silkworm and C. elegans, respectively. To get insights on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service