Skip to Content
Merck
All Photos(2)

Documents

HPA044511

Sigma-Aldrich

Anti-CXXC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CFP1, Anti-CGBP, Anti-CXXC Finger Protein 1, Anti-HsT2645, Anti-PCCX1, Anti-PHF18, Anti-SPP1, Anti-ZCGPC1, Anti-hCGBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VKVKHVKRREKKSEKKKEERYKRHRQKQKHKDKWKHPERADAKDPASLPQCLGPGCVRPAQPSSKYCSDDCGMKLAANRIYEILPQRIQQWQQSPCIAEEHGKKLLERIRREQQSARTRLQEMERRFHELEAIILRAKQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXXC1(30827)

General description

The gene CXXC1 (CXXC finger protein 1) is mapped to human chromosome 18q21. The protein has two plant homeodomains, a cysteine-rich CXXC DNA-binding domain, acidic, basic, and coiled-coil domains and a Set1 interaction domain (SID). It is present with euchromatic regions of the genome.

Immunogen

CXXC finger protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CXXC1 (CXXC finger protein 1) is a non-enzymatic component of the COMPASS (complex of proteins associated with set1) histone methyltransferase complex. It mainly interacts with non-methylated CpG (cytosine–guanosine) motifs via CXXC domain and particularly prefers a CpGG motif. CXXC1 epigenetically regulates cytosine and histone methylation. It is important for vertebrate development and absence of CXXC1 causes embryonic lethality. Low levels of CXXC1 in embryonic stem cells causes 70% reduction in cytosine methylation and 60% less DNA methyltransferase (DNMT1) activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70946

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pilot Study for Discovering Candidate Genes of Chromosome 18q21 in Methamphetamine Abusers: Case-control Association Study.
Lee BD, et al.
Clinical psychopharmacology and neuroscience : the official scientific journal of the Korean College of Neuropsychopharmacology, 12, 54-64 (2014)
CXXC finger protein 1 contains redundant functional domains that support embryonic stem cell cytosine methylation, histone methylation, and differentiation.
Tate CM, et al.
Molecular and Cellular Biology, 29, 3817-3831 (2009)
CFP1 interacts with DNMT1 independently of association with the Setd1 Histone H3K4 methyltransferase complexes.
Butler JS, et al.
Dna and Cell Biology, 27, 533-543 (2008)
The structural basis for selective binding of non-methylated CpG islands by the CFP1 CXXC domain.
Xu C, et al.
Nature Communications, 2, 227-227 (2011)
Induction of activation-induced cytidine deaminase-targeting adaptor 14-3-3? is mediated by NF-?B-dependent recruitment of CFP1 to the 5'-CpG-3'-rich 14-3-3? promoter and is sustained by E2A.
Mai T, et al.
Journal of Immunology, 191, 1895-1906 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service