Skip to Content
Merck
All Photos(9)

Key Documents

HPA037559

Sigma-Aldrich

Anti-MKRN2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSPC070, Anti-Makorin ring finger protein 2, Anti-RNF62

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human, rat, mouse
citations:
3
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NSHEPGKREKRTLVLRDRNLSGMAERKTQPSMVSNPGSCSDPQPSPEMKPHSYLDAIRSGLDDVEASSSYSNEQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKRN2(23609)

General description

Makorin-2 (MKRN2) is a novel ubiquitin E3 ligase that is also known as HSPC070. It is a member of the MKRN gene family. It has four C3H zinc fingers and a signature C3HC4 RING zinc finger domain. MKRN2 is present in the cytoplasm and the nucleus. This gene is located on human chromosome 3p25.

Immunogen

makorin ring finger protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Makorin-2 (MKRN2) induces ubiquitination and degradation of p65 with the help of RING finger domain. It negatively modulates inflammatory responses.MKRN2 helps to transport p65 from soluble to insoluble nuclear compartments.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79310

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Jiang et al.
Journal of experimental & clinical cancer research : CR, 37(1), 189-189 (2018-08-15)
Makorin RING zinc finger-2 (MKRN2) belongs to the makorin RING zinc finger family and is a novel ubiquitin E3 ligase targeting the p65 subunit of NF-κB to negatively regulate inflammatory responses; however, the relationship between MKRN2 and tumorigenesis remains unclear.
Chanyoung Shin et al.
Scientific reports, 7, 46097-46097 (2017-04-06)
Activation of NF-κB transcription factor is strictly regulated to prevent excessive inflammatory responses leading to immunopathology. However, it still remains unclear how NF-κB activation is negatively controlled. The PDZ-LIM domain-containing protein PDLIM2 is a nuclear ubiquitin E3 ligase targeting the
Ubiquitous expression of MAKORIN-2 in normal and malignant hematopoietic cells and its growth promoting activity.
Lee KY, et al.
PLoS ONE, 9(3), e92706-e92706 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service