Skip to Content
Merck
All Photos(6)

Key Documents

HPA030200

Sigma-Aldrich

Anti-DST antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BP240, Anti-BPA, Anti-BPAG1, Anti-CATX-15, Anti-FLJ13425, Anti-FLJ21489, Anti-FLJ30627, Anti-FLJ32235, Anti-KIAA0728, Anti-MACF2, Anti-dystonin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on30 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KPRCQNVAEDMDIQLCASILNENSDENENINTMILLDKMHSCSSLEKQQRVNVVQLASPSENNLVTEKSNLPEYTTEIAGKSKENLLNHE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DST(667)

General description

DST gene encodes dystonin, a cytoskeletal linker protein. Dystonin proteins are present as several isoforms in neural and muscle cells. DST gene is located on the human chromosome 6p12.1. Dystonin protein encompasses plakin and spectrin domains.

Immunogen

dystonin recombinant protein epitope signature tag (PrEST)

Application

Anti-DST antibody produced in rabbit is suitable for human dystonin detection in live cell microscopy.

Biochem/physiol Actions

DST integrates microtubules, actin and intermediate filaments and is essential for cytoskeleton network in neuronal cells. A frame shift mutation in the DST gene affects sensory and autonomous nervous system, leading to hereditary sensory autonomic neuropathy type VI. The incidence of nonsense mutation in the DST gene is implicated in the skin disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77625

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel mutations in dystonin provide clues to the pathomechanisms of HSAN-VI
Manganelli F, et al.
Neurology, 88(22), 2132-2140 (2017)
Hereditary sensory autonomic neuropathy caused by a mutation in dystonin
Edvardson S, et al.
Annals of Neurology, 71(4), 569-572 (2012)
Structural analysis of the plakin domain of bullous pemphigoid antigen1 (BPAG1) suggests that plakins are members of the spectrin superfamily
Jefferson JJ, et al.
Journal of Molecular Biology, 366(1), 244-257 (2007)
A homozygous nonsense mutation within the dystonin gene coding for the coiled-coil domain of the epithelial isoform of BPAG1 underlies a new subtype of autosomal recessive epidermolysis bullosa simplex
Groves RW, et al.
The Journal of Investigative Dermatology, 130(6), 1551-1557 (2010)
Cloning and characterization of the neural isoforms of human dystonin
Brown A, et al.
Genomics, 29(3), 777-780 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service