Skip to Content
Merck
All Photos(6)

Key Documents

HPA029816

Sigma-Aldrich

Anti-GADD45B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARC21, Anti-DKFZP566B133, Anti-GADD45β, Anti-MYD118, Anti-OTTHUMG00000008026, Anti-growth arrest and DNA-damage-inducible, β, Anti-p21-Arc

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AVEELLVAAQRQDRLTVGVYESAKLMNVDPDSVVLCLLAIDEEEEDDIALQIHFTLIQSFCCDNDINIVRVSGMQRLAQLLGEPAETQGTTEARDLHCLLVTNPHTDAWKSHGLVEVASYCEESRGNNQWVPY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GADD45B(4616)

General description

Growth arrest and DNA damage-inducible-β (Gadd45β) protein belongs to the GADD45 family. It forms a homodimer or heterodimer with other proteins from Gadd45 family.[1] The Gadd45β gene is mapped to human chromosome19p13.3.

Immunogen

growth arrest and DNA-damage-inducible, beta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-GADD45B antibody produced in rabbit has been used in immunohistochemical (IHC) staining at a dilution of 1:500.[1]

Biochem/physiol Actions

Growth arrest and DNA damage-inducible-β (Gadd45β) is a stress-response protein that is expressed in response to environmental or physiological stress. Its abnormal expression is implicated in tumor development.[1] GADD45β interacts with p38 mitogen-activated protein kinase (MAPK) and acts as an inhibitor of mitogen-activated protein kinase kinases (MKK7/MKK4). Deficiency of GADD45β may lead to pronounced severity in allergic encephalomyelitis, drug-induced liver injury, and hepatocellular carcinoma in murine models. GADD45β is activated in murine myeloid leukemia cells. It may serve as a biomarker for myocardium-associated hypoxia-induced apoptosis in newborn infants.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72472

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung-Je Moon et al.
Journal of cellular physiology, 233(10), 7128-7138 (2018-05-10)
Sepsis is a systemic inflammatory response syndrome due to microbial infection. Growth arrest and DNA-damage-inducible 45 beta (GADD45β) are induced by genotoxic stress and inflammatory cytokines. However, the role of GADD45β during bacterial infection remains unclear. This study was aimed at
Mona Adalgisa Simões et al.
Pathology, research and practice, 213(3), 222-226 (2017-02-19)
Among the several organs affected by perinatal hypoxia, the heart plays a central role, with cell death caused mainly by apoptosis. One of the biomarkers most often linked to hypoxia-derived apoptosis of cardiomyocytes in animals is Gadd45β. From the published
Kyaw Zwar Myint et al.
Oncology letters, 15(3), 3031-3041 (2018-02-13)
Growth arrest and DNA damage-inducible-β (Gadd45β) is a stress-response protein involved in a number of processes, including cell cycle control, DNA repair, survival and death control, and stress signaling, depending on its interactions. Gadd45β expression is dysregulated in numerous types
Tao Yang et al.
International orthopaedics, 36(6), 1307-1314 (2011-12-21)
To investigate the effects of genetic factors on idiopathic scoliosis (IS) and genetic modes through genetic epidemiological survey on IS in Chongqing City, China, and to determine whether SH3GL1, GADD45B, and FGF22 in the chromosome 19p13.3 are the pathogenic genes
Anthony E Rizzardi et al.
BMC cancer, 14, 244-244 (2014-04-09)
Prognostic multibiomarker signatures in prostate cancer (PCa) may improve patient management and provide a bridge for developing novel therapeutics and imaging methods. Our objective was to evaluate the association between expression of 33 candidate protein biomarkers and time to biochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service